Skip to Content
Merck
All Photos(1)

Key Documents

COM448622273

Combi-Blocks, Inc.

5-Bromo-6,7-difluoro-1-methylbenzotriazole

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 392.28
5 G
CHF 1’222.98

Vendor SKU: IN-0391-1G

CHF 392.28


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 392.28
5 G
CHF 1’222.98

About This Item

CAS Number:
Molecular Weight:
248.03
MDL number:

Vendor SKU: IN-0391-1G

CHF 392.28


Ships in 14 days - from Aldrich Partner.

Iupac Name

5-bromo-6,7-difluoro-1-methyl-1H-1,2,3-benzotriazole

Inchi Code

1S/C7H4BrF2N3/c1-13-7-4(11-12-13)2-3(8)5(9)6(7)10/h2H,1H3

InChI key

YHCPPQCLQCMBSO-UHFFFAOYSA-N

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?IN-0391

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?IN-0391


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service