Skip to Content
Merck
All Photos(1)

Key Documents

COM448622218

Combi-Blocks, Inc.

1-Cyclohexyl-6-fluoro-1,3-benzodiazole

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 170.76
5 G
CHF 392.28
25 G
CHF 1’001.46

Vendor SKU: IN-0319-1G

CHF 170.76


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 170.76
5 G
CHF 392.28
25 G
CHF 1’001.46

About This Item

CAS Number:
Molecular Weight:
218.27
MDL number:

Vendor SKU: IN-0319-1G

CHF 170.76


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-cyclohexyl-6-fluoro-1H-benzimidazole

Inchi Code

1S/C13H15FN2/c14-10-6-7-12-13(8-10)16(9-15-12)11-4-2-1-3-5-11/h6-9,11H,1-5H2

InChI key

VNVCGBCUQSDFOA-UHFFFAOYSA-N

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?IN-0319

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?IN-0319


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service