Skip to Content
Merck
All Photos(1)

Key Documents

COM448617062

Combi-Blocks, Inc.

4-Methoxybenzyl 5-bromo-2-chlorobenzamide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 156.92
5 G
CHF 309.21
25 G
CHF 724.56

Vendor SKU: CA-4725-1G

CHF 156.92


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 156.92
5 G
CHF 309.21
25 G
CHF 724.56

About This Item

CAS Number:
Molecular Weight:
354.63
MDL number:

Vendor SKU: CA-4725-1G

CHF 156.92


Ships in 14 days - from Aldrich Partner.

Iupac Name

5-bromo-2-chloro-N-(4-methoxybenzyl)benzamide

Inchi Code

1S/C15H13BrClNO2/c1-20-12-5-2-10(3-6-12)9-18-15(19)13-8-11(16)4-7-14(13)17/h2-8H,9H2,1H3,(H,18,19)

InChI key

UGBPFPJQRDBYHO-UHFFFAOYSA-N

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?CA-4725

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?CA-4725


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service