CIAH98B92D0B
ChemScene LLC (Preferred Partner)
(S)-4-Ethynyl-2,2-dimethyl-1,3-dioxolane
About This Item
bp
129.6+-35.0 C at 760 mmHg
Iupac Name
(4S)-4-ethynyl-2,2-dimethyl-1,3-dioxolane
Inchi Code
1S/C7H10O2/c1-4-6-5-8-7(2,3)9-6/h1,6H,5H2,2-3H3/t6-/m0/s1
InChI key
GPNBPQQQZHDRMZ-LURJTMIESA-N
storage temp.
Purity
Country of Origin
Physical Form
Product Link
Boiling point
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0433216.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service