Skip to Content
Merck
All Photos(1)

Key Documents

CH6947294146

Chem Impex International, Inc.

4-[(Boc)2-guanidino]phenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
393.44
MDL number:

mp

130 - 131 C

Iupac Name

[4-({bis[(tert-butoxycarbonyl)amino]methylene}amino)phenyl]acetic acid

Inchi Code

1S/C19H27N3O6/c1-18(2,3)27-16(25)21-15(22-17(26)28-19(4,5)6)20-13-9-7-12(8-10-13)11-14(23)24/h7-10H,11H2,1-6H3,(H,23,24)(H2,20,21,22,25,26)

InChI key

NSLPHFNMIHNLKV-UHFFFAOYSA-N

storage temp.

Store at 0 - 8 C

Purity

99%

Shipping Temperature

RT

Physical Form

Light yellow crystalline powder

Product Link

https://www.chemimpex.com/product/productinfo/4-boc-sub-2-sub-guanidino-phenylacetic-acid/20896?cid=1

Melting Point

130 - 131 C

MSDS

https://www.chemimpex.com/product/productinfo/4-boc-sub-2-sub-guanidino-phenylacetic-acid/20896?cid=1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service