Skip to Content
Merck
All Photos(1)

Key Documents

C12H316CF12B

Chemcia Scientific, LLC

6-[Bis(tert-Butoxycarbonyl)amino]-nicotinonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 264.48
1 G
CHF 360.33
5 G
CHF 1’146.30

Vendor SKU: CC00-9609-500MG

CHF 264.48


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
500 MG
CHF 264.48
1 G
CHF 360.33
5 G
CHF 1’146.30

About This Item

CAS Number:
Molecular Weight:
319.36
MDL number:

Vendor SKU: CC00-9609-500MG

CHF 264.48


Ships in 14 days - from Aldrich Partner.

Iupac Name

tert-butyl (tert-butoxycarbonyl)(5-cyanopyridin-2-yl)carbamate

Inchi Code

1S/C16H21N3O4/c1-15(2,3)22-13(20)19(14(21)23-16(4,5)6)12-8-7-11(9-17)10-18-12/h7-8,10H,1-6H3

InChI key

NRNUFXYWTMKOAL-UHFFFAOYSA-N

storage temp.

0-5C

Purity

97%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

White Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=6729

MSDS

http://www.chemcia.com/product_fl02.asp?id=6729


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service