Skip to Content
Merck
All Photos(1)

Key Documents

CH4238556539

ChemBridge Corporation

5-(3-hydroxy-3-methylbut-1-yn-1-yl)nicotinic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 320.71
5 G
CHF 1’100.29

Vendor SKU: 3002769-1G

CHF 320.71


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 320.71
5 G
CHF 1’100.29

About This Item

CAS Number:
Molecular Weight:
205.21
MDL number:

Vendor SKU: 3002769-1G

CHF 320.71


Ships in 14 days - from Aldrich Partner.

Iupac Name

5-(3-hydroxy-3-methyl-1-butynyl)nicotinic acid

Inchi Code

1S/C11H11NO3/c1-11(2,15)4-3-8-5-9(10(13)14)7-12-6-8/h5-7,15H,1-2H3,(H,13,14)

InChI key

YHPRXKWSTJOFBT-UHFFFAOYSA-N

Purity

95%

Country of Origin

KZ

Product Link

https://www.hit2lead.com/building-blocks/3002769

MSDS

https://www.hit2lead.com/sds/3002769_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service