Skip to Content
Merck
All Photos(1)

Key Documents

CH4182076499

ChemBridge Corporation

N-[(1-ethylpyrrolidin-3-yl)methyl]ethanamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 91.95
5 G
CHF 163.52
10 G
CHF 228.70
25 G
CHF 384.61
50 G
CHF 577.59

Vendor SKU: 4026486-1G

CHF 91.95


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 91.95
5 G
CHF 163.52
10 G
CHF 228.70
25 G
CHF 384.61
50 G
CHF 577.59

About This Item

CAS Number:
Molecular Weight:
156.27
MDL number:

Vendor SKU: 4026486-1G

CHF 91.95


Ships in 14 days - from Aldrich Partner.

Iupac Name

N-[(1-ethyl-3-pyrrolidinyl)methyl]ethanamine

Inchi Code

1S/C9H20N2/c1-3-10-7-9-5-6-11(4-2)8-9/h9-10H,3-8H2,1-2H3

InChI key

JMAGSGRQNWLQKT-UHFFFAOYSA-N

storage temp.

RT

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

liquid

Product Link

https://www.hit2lead.com/building-blocks/4026486

MSDS

https://www.hit2lead.com/sds/4026486_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service