Skip to Content
Merck
All Photos(1)

Key Documents

CH4182074895

ChemBridge Corporation

3-propylmorpholine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 149.46
1 G
CHF 198.02
5 G
CHF 610.82

Vendor SKU: 4021666-250MG

CHF 149.46


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CHF 149.46
1 G
CHF 198.02
5 G
CHF 610.82

About This Item

CAS Number:
Molecular Weight:
129.2
MDL number:

Vendor SKU: 4021666-250MG

CHF 149.46


Ships in 14 days - from Aldrich Partner.

Iupac Name

3-propylmorpholine

Inchi Code

1S/C7H15NO/c1-2-3-7-6-9-5-4-8-7/h7-8H,2-6H2,1H3

InChI key

ACDQSRFDVDVZRK-UHFFFAOYSA-N

storage temp.

RT

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

liquid

Product Link

https://www.hit2lead.com/building-blocks/4021666

MSDS

https://www.hit2lead.com/sds/4021666_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service