Skip to Content
Merck
All Photos(1)

Key Documents

BL3H160C4A43

BLD Pharmatech Co., Ltd.

2-(2-(2-Methoxyethoxy)ethoxy)ethyl methanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 60.70
250 MG
CHF 87.15
1 G
CHF 141.20

Vendor SKU: BD760170-100MG

CHF 60.70


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 60.70
250 MG
CHF 87.15
1 G
CHF 141.20

About This Item

CAS Number:
Molecular Weight:
242.29
MDL number:

Vendor SKU: BD760170-100MG

CHF 60.70


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-(2-(2-methoxyethoxy)ethoxy)ethyl methanesulfonate

Inchi Code

1S/C8H18O6S/c1-11-3-4-12-5-6-13-7-8-14-15(2,9)10/h3-8H2,1-2H3

InChI key

ZMRMMAUVJFLPIV-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Room Temperature

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Colorless to Yellow Liquid or Semi-Solid or solid

Product Link

http://www.bldpharm.com/products/74654-05-0.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://file.bldpharm.com/static/upload/prosds/bldsds/761/SDS-BD760170.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service