Skip to Content
Merck
All Photos(1)

Key Documents

ATC456776887

Atlantic Research Chemicals Ltd

3-Bromo-2-oxopropanoyl chloride (as a 50% soln in DCM)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 275.20
500 MG
CHF 396.16
1 G
CHF 510.40
2.5 G
CHF 1’014.40

Vendor SKU: CC10115-250MG

CHF 275.20


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
250 MG
CHF 275.20
500 MG
CHF 396.16
1 G
CHF 510.40
2.5 G
CHF 1’014.40

About This Item

CAS Number:
Molecular Weight:
185.4
MDL number:

Vendor SKU: CC10115-250MG

CHF 275.20


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

3-bromo-2-oxopropanoyl chloride

Inchi Code

1S/C3H2BrClO2/c4-1-2(6)3(5)7/h1H2

InChI key

UMNDXFOJQBVTOP-UHFFFAOYSA-N

Purity

90

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=CC10115

MSDS

https://atlantic-chemicals.com/product/?code=CC10115


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service