Skip to Content
Merck
All Photos(1)

Key Documents

ATC311762356

Atlantic Research Chemicals Ltd

5-Chloro-2-(4-chlorophenoxy)aniline

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 62.85
1 G
CHF 63.52
5 G
CHF 83.68

Vendor SKU: AE00153-250MG

CHF 62.85


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CHF 62.85
1 G
CHF 63.52
5 G
CHF 83.68

About This Item

CAS Number:
Molecular Weight:
254.11
MDL number:

Vendor SKU: AE00153-250MG

CHF 62.85


Ships in 14 days - from Aldrich Partner.

Iupac Name

5-chloro-2-(4-chlorophenoxy)aniline

Inchi Code

1S/C12H9Cl2NO/c13-8-1-4-10(5-2-8)16-12-6-3-9(14)7-11(12)15/h1-7H,15H2

InChI key

WLJSUJOESWTGEX-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=AE00153

MSDS

https://atlantic-chemicals.com/product/?code=AE00153


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service