Skip to Content
Merck
All Photos(1)

Key Documents

ATEH996C54B0

AstaTech, Inc.

1-FLUORO-4-(METHYLSULFINYL)BENZENE

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 177.15
1 G
CHF 276.99

Vendor SKU: W16289-250MG

CHF 177.15


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CHF 177.15
1 G
CHF 276.99

About This Item

CAS Number:
Molecular Weight:
158.2
MDL number:

Vendor SKU: W16289-250MG

CHF 177.15


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-fluoro-4-(methylsulfinyl)benzene

Inchi Code

1S/C7H7FOS/c1-10(9)7-4-2-6(8)3-5-7/h2-5H,1H3

InChI key

PQOHTVHAGPPTEE-UHFFFAOYSA-N

Purity

95%

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=125314

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W16289


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service