Skip to Content
Merck
All Photos(1)

Key Documents

ATEH96F1258E

AstaTech, Inc.

METHYL 4,5,6,7-TETRAHYDRO-1H-BENZO[D]IMIDAZOLE-6-CARBOXYLATE

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 442.07
250 MG
CHF 769.56
1 G
CHF 2’055.54

Vendor SKU: D75447-100MG

CHF 442.07


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 442.07
250 MG
CHF 769.56
1 G
CHF 2’055.54

About This Item

CAS Number:
Molecular Weight:
180.21
MDL number:

Vendor SKU: D75447-100MG

CHF 442.07


Ships in 14 days - from Aldrich Partner.

Iupac Name

methyl 4,5,6,7-tetrahydro-1H-benzimidazole-6-carboxylate

Inchi Code

1S/C9H12N2O2/c1-13-9(12)6-2-3-7-8(4-6)11-5-10-7/h5-6H,2-4H2,1H3,(H,10,11)

InChI key

MJLAMGHGLJXZLT-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=188500

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=D75447


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service