Skip to Content
Merck
All Photos(1)

Key Documents

ATEH95EB7788

AstaTech, Inc.

6-BROMO-9H-PYRIDO[3,4-B]INDOLE

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 358.20
50 MG
CHF 954.60

Vendor SKU: D84363-10MG

CHF 358.20


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
10 MG
CHF 358.20
50 MG
CHF 954.60

About This Item

CAS Number:
Molecular Weight:
247.09
MDL number:

Vendor SKU: D84363-10MG

CHF 358.20


Ships in 14 days - from Aldrich Partner.

Iupac Name

6-bromo-9H-beta-carboline

Inchi Code

1S/C11H7BrN2/c12-7-1-2-10-9(5-7)8-3-4-13-6-11(8)14-10/h1-6,14H

InChI key

DNUXGDOJLQYUTO-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=204783

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=D84363


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service