Skip to Content
Merck
All Photos(1)

Key Documents

ATEH57C9A6E4

AstaTech, Inc.

4-METHYL-5-NITRO-2,3'-BIPYRIDINE

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 370.18
250 MG
CHF 961.26
1 G
CHF 2’691.88

Vendor SKU: C90195-50MG

CHF 370.18


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CHF 370.18
250 MG
CHF 961.26
1 G
CHF 2’691.88

About This Item

CAS Number:
Molecular Weight:
215.21
MDL number:

Vendor SKU: C90195-50MG

CHF 370.18


Ships in 14 days - from Aldrich Partner.

Iupac Name

4-methyl-5-nitro-2,3'-bipyridine

Inchi Code

1S/C11H9N3O2/c1-8-5-10(9-3-2-4-12-6-9)13-7-11(8)14(15)16/h2-7H,1H3

InChI key

WMRKLFBOOLQJKN-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=166846

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C90195


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service