Skip to Content
Merck
All Photos(1)

Key Documents

ATE959437699

AstaTech, Inc.

TERT-BUTYL ((4-CYANOPHENYL)(PHENYLSULFONYL)METHYL)CARBAMATE

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 402.13
1 G
CHF 918.66
5 G
CHF 2’634.64

Vendor SKU: P16913-250MG

CHF 402.13


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
250 MG
CHF 402.13
1 G
CHF 918.66
5 G
CHF 2’634.64

About This Item

CAS Number:
Molecular Weight:
372.44
MDL number:

Vendor SKU: P16913-250MG

CHF 402.13


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

tert-butyl ((4-cyanophenyl)(phenylsulfonyl)methyl)carbamate

Inchi Code

1S/C19H20N2O4S/c1-19(2,3)25-18(22)21-17(15-11-9-14(13-20)10-12-15)26(23,24)16-7-5-4-6-8-16/h4-12,17H,1-3H3,(H,21,22)

InChI key

WRSNHXTZVJGPKK-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=102501

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=P16913


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service