Skip to Content
Merck

ATE514315952

AstaTech, Inc.

4-(4-(OXETAN-3-YL)PIPERAZIN-1-YL)ANILINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
233.31
MDL number:

Pricing and availability is not currently available.

Iupac Name

4-(4-(oxetan-3-yl)piperazin-1-yl)aniline

Inchi Code

1S/C13H19N3O/c14-11-1-3-12(4-2-11)15-5-7-16(8-6-15)13-9-17-10-13/h1-4,13H,5-10,14H2

InChI key

NSMYDEYAHKTAGB-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=80503

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W11307


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service