Skip to Content
Merck
All Photos(1)

Key Documents

ATE514314996

AstaTech, Inc.

2-(PYRIDIN-3-YLMETHYL)QUINUCLIDIN-3-ONE

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 348.88
250 MG
CHF 591.17
1 G
CHF 1’347.32

Vendor SKU: W10352-100MG

CHF 348.88


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 348.88
250 MG
CHF 591.17
1 G
CHF 1’347.32

About This Item

CAS Number:
Molecular Weight:
216.28
MDL number:

Vendor SKU: W10352-100MG

CHF 348.88


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-(pyridin-3-ylmethyl)quinuclidin-3-one

Inchi Code

1S/C13H16N2O/c16-13-11-3-6-15(7-4-11)12(13)8-10-2-1-5-14-9-10/h1-2,5,9,11-12H,3-4,6-8H2

InChI key

RDZRUHBKCNPOPR-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=79563

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W10352


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service