Skip to Content
Merck
All Photos(1)

Key Documents

ATE443547888

AstaTech, Inc.

TERT-BUTYL 4-FLUORO-3-OXOAZEPANE-1-CARBOXYLATE

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 311.61
250 MG
CHF 394.14

Vendor SKU: 73064-100MG

CHF 311.61


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 311.61
250 MG
CHF 394.14

About This Item

CAS Number:
Molecular Weight:
231.27
MDL number:

Vendor SKU: 73064-100MG

CHF 311.61


Ships in 14 days - from Aldrich Partner.

Iupac Name

tert-butyl 4-fluoro-3-oxoazepane-1-carboxylate

Inchi Code

1S/C11H18FNO3/c1-11(2,3)16-10(15)13-6-4-5-8(12)9(14)7-13/h8H,4-7H2,1-3H3

InChI key

WIJMFJCEVOJLQM-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=56094

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=73064


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service