Skip to Content
Merck
All Photos(1)

Key Documents

ATE423889671

AstaTech, Inc.

2-AMINO-2-(PYRROLIDIN-3-YL)ACETIC ACID 2HCL

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 291.64
1 G
CHF 671.04
5 G
CHF 2’026.26

Vendor SKU: 67423-250MG

CHF 291.64


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CHF 291.64
1 G
CHF 671.04
5 G
CHF 2’026.26

About This Item

CAS Number:
Molecular Weight:
217.09
MDL number:

Vendor SKU: 67423-250MG

CHF 291.64


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-amino-2-(pyrrolidin-3-yl)acetic acid dihydrochloride

Inchi Code

1S/C6H12N2O2.2ClH/c7-5(6(9)10)4-1-2-8-3-4;;/h4-5,8H,1-3,7H2,(H,9,10);2*1H

InChI key

HOBJVOBXDWYSLG-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=9172

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=67423


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service