Skip to Content
Merck
All Photos(1)

Key Documents

ATE398811679

AstaTech, Inc.

(R)-1-BENZYL 3-ETHYL PIPERIDINE-1,3-DICARBOXYLATE

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 162.51
1 G
CHF 295.63
5 G
CHF 774.88

Vendor SKU: 81469-250MG

CHF 162.51


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CHF 162.51
1 G
CHF 295.63
5 G
CHF 774.88

About This Item

CAS Number:
Molecular Weight:
291.35
MDL number:

Vendor SKU: 81469-250MG

CHF 162.51


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-benzyl 3-ethyl (3R)-1,3-piperidinedicarboxylate

Inchi Code

1S/C16H21NO4/c1-2-20-15(18)14-9-6-10-17(11-14)16(19)21-12-13-7-4-3-5-8-13/h3-5,7-8,14H,2,6,9-12H2,1H3/t14-/m1/s1

InChI key

NJRONXOELMBCSE-CQSZACIVSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=18068

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=81469


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service