Skip to Content
Merck
All Photos(1)

Key Documents

ATE245844554

AstaTech, Inc.

ETHYL 2-MORPHOLINOISONICOTINATE

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 265.01
5 G
CHF 772.22

Vendor SKU: 30046-1G

CHF 265.01


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 265.01
5 G
CHF 772.22

About This Item

CAS Number:
Molecular Weight:
236.27
MDL number:

Vendor SKU: 30046-1G

CHF 265.01


Ships in 14 days - from Aldrich Partner.

Iupac Name

ethyl 2-(4-morpholinyl)isonicotinate

Inchi Code

1S/C12H16N2O3/c1-2-17-12(15)10-3-4-13-11(9-10)14-5-7-16-8-6-14/h3-4,9H,2,5-8H2,1H3

InChI key

UBQGLKBSCMRITK-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

97%

Shipping Temperature

Normal

Product Link

http://astatechinc.com/CPSResult.php?CRNO=1045

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=30046


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service