Skip to Content
Merck
All Photos(1)

Key Documents

ATE245844368

AstaTech, Inc.

4-METHYL-PYRIDINE-2-SULFONYL CHLORIDE

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 375.51
250 MG
CHF 668.38
1 G
CHF 1’387.26

Vendor SKU: 27044-100MG

CHF 375.51


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CHF 375.51
250 MG
CHF 668.38
1 G
CHF 1’387.26

About This Item

CAS Number:
Molecular Weight:
191.64
MDL number:

Vendor SKU: 27044-100MG

CHF 375.51


Ships in 14 days - from Aldrich Partner.

Iupac Name

4-methyl-2-pyridinesulfonyl chloride

Inchi Code

1S/C6H6ClNO2S/c1-5-2-3-8-6(4-5)11(7,9)10/h2-4H,1H3

InChI key

YIQOZBGOBMKGRZ-UHFFFAOYSA-N

storage temp.

-20 C

Purity

97%

Country of Origin

CN

Shipping Temperature

Cold box

Physical Form

Pale-yellow to Yellow-brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=830

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=27044


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service