Skip to Content
Merck
All Photos(1)

Key Documents

APOH57C99DA0

Apollo Scientific Ltd

2,3-Difluoro-4-(trifluoromethoxy)aniline

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 225.90
1 G
CHF 598.80
5 G
CHF 1’717.50

Vendor SKU: PC51845-250MG

CHF 225.90


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CHF 225.90
1 G
CHF 598.80
5 G
CHF 1’717.50

About This Item

CAS Number:
Molecular Weight:
213.11
MDL number:

Vendor SKU: PC51845-250MG

CHF 225.90


Ships in 14 days - from Aldrich Partner.

Iupac Name

2,3-difluoro-4-(trifluoromethoxy)aniline

Inchi Code

1S/C7H4F5NO/c8-5-3(13)1-2-4(6(5)9)14-7(10,11)12/h1-2H,13H2

InChI key

SLBZPAVVVANJBN-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid-Oil

Product Link

https://store.apolloscientific.co.uk/product/23-difluoro-4-trifluoromethoxyaniline

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/23-difluoro-4-trifluoromethoxyaniline


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service