Skip to Content
Merck
All Photos(1)

Key Documents

APO715372463

Apollo Scientific Ltd

3-Iodo-4-(trifluoromethyl)benzonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 135.70
1 G
CHF 381.00

Vendor SKU: PC502103-250MG

CHF 135.70


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
250 MG
CHF 135.70
1 G
CHF 381.00

About This Item

CAS Number:
Molecular Weight:
297.02
MDL number:

Vendor SKU: PC502103-250MG

CHF 135.70


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

3-iodo-4-(trifluoromethyl)benzonitrile

Inchi Code

1S/C8H3F3IN/c9-8(10,11)6-2-1-5(4-13)3-7(6)12/h1-3H

InChI key

FXIYYECGGVTKBN-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/3-iodo-4-trifluoromethylbenzonitrile

Pictograms

Signal Word

Danger

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-iodo-4-trifluoromethylbenzonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service