Skip to Content
Merck
All Photos(1)

Key Documents

APO456876923

Apollo Scientific Ltd

2,3,6-Trifluorophenylacetonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 59.80
5 G
CHF 99.40
25 G
CHF 286.40

Vendor SKU: PC7818L-1G

CHF 59.80


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 59.80
5 G
CHF 99.40
25 G
CHF 286.40

About This Item

CAS Number:
Molecular Weight:
171.12
MDL number:

Vendor SKU: PC7818L-1G

CHF 59.80


Ships in 14 days - from Aldrich Partner.

Iupac Name

(2,3,6-trifluorophenyl)acetonitrile

Inchi Code

1S/C8H4F3N/c9-6-1-2-7(10)8(11)5(6)3-4-12/h1-2H,3H2

InChI key

PQIFHBOOYBTJLD-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/236-trifluorophenylacetonitrile

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/236-trifluorophenylacetonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service