Skip to Content
Merck
All Photos(1)

Key Documents

APO456790325

Apollo Scientific Ltd

2-(Methylamino)nicotinonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 88.40

Vendor SKU: OR12119-1G

CHF 88.40


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 88.40

About This Item

CAS Number:
Molecular Weight:
133.15
MDL number:

Vendor SKU: OR12119-1G

CHF 88.40


Ships in 14 days - from Aldrich Partner.

bp

85-87

Iupac Name

2-(methylamino)nicotinonitrile

Inchi Code

1S/C7H7N3/c1-9-7-6(5-8)3-2-4-10-7/h2-4H,1H3,(H,9,10)

InChI key

AGZQPTBBDDQFKJ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/2-methylaminonicotinonitrile

Boiling point

85-87

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-methylaminonicotinonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service