Skip to Content
Merck
All Photos(1)

Key Documents

APO455837641

Apollo Scientific Ltd

4-Bromo-2-chloro-5-fluorobenzoyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 96.10
1 G
CHF 231.40

Vendor SKU: PC6589-250MG

CHF 96.10


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CHF 96.10
1 G
CHF 231.40

About This Item

CAS Number:
Molecular Weight:
271.9
MDL number:

Vendor SKU: PC6589-250MG

CHF 96.10


Ships in 14 days - from Aldrich Partner.

Iupac Name

4-bromo-2-chloro-5-fluorobenzoyl chloride

Inchi Code

1S/C7H2BrCl2FO/c8-4-2-5(9)3(7(10)12)1-6(4)11/h1-2H

InChI key

IQFUAYKLNRHXJT-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/4-bromo-2-chloro-5-fluorobenzoyl-chloride

Pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-bromo-2-chloro-5-fluorobenzoyl-chloride


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service