Skip to Content
Merck
All Photos(1)

Key Documents

APO455836421

Apollo Scientific Ltd

O-[3-(Trifluoromethyl)benzyl]hydroxylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
227.61
MDL number:

bp

160-170

Iupac Name

1-[(aminooxy)methyl]-3-(trifluoromethyl)benzene hydrochloride

Inchi Code

1S/C8H8F3NO.ClH/c9-8(10,11)7-3-1-2-6(4-7)5-13-12;/h1-4H,5,12H2;1H

InChI key

DXEDFEUKJRKFOV-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/o-3-trifluoromethylbenzylhydroxylamine-hydrochloride

Synonym

3-[(Aminooxy)methyl]benzotrifluoride hydrochloride, 1-[(Aminooxy)methyl]-3-(trifluoromethyl)benzene hydrochloride

Boiling point

160-170

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/o-3-trifluoromethylbenzylhydroxylamine-hydrochloride


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service