Skip to Content
Merck
All Photos(1)

Key Documents

APO455835325

Apollo Scientific Ltd

2-Amino-4-(trifluoromethoxy)phenol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
193.13
MDL number:

bp

98-101

Iupac Name

2-amino-4-(trifluoromethoxy)phenol

Inchi Code

1S/C7H6F3NO2/c8-7(9,10)13-4-1-2-6(12)5(11)3-4/h1-3,12H,11H2

InChI key

DNYJUUSAYDJKAF-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/2-amino-4-trifluoromethoxyphenol

Synonym

2-Hydroxy-5-(trifluoromethoxy)aniline

Boiling point

98-101

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-amino-4-trifluoromethoxyphenol


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service