Skip to Content
Merck
All Photos(1)

Key Documents

APO455833961

Apollo Scientific Ltd

5-Fluoro-2-methylbenzylamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 66.40
5 G
CHF 130.20
25 G
CHF 381.00

Vendor SKU: PC0359-1G

CHF 66.40


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 66.40
5 G
CHF 130.20
25 G
CHF 381.00

About This Item

CAS Number:
Molecular Weight:
139.17
MDL number:

Vendor SKU: PC0359-1G

CHF 66.40


Ships in 14 days - from Aldrich Partner.

Iupac Name

5-fluoro-2-methylbenzylamine

Inchi Code

1S/C8H10FN/c1-6-2-3-8(9)4-7(6)5-10/h2-4H,5,10H2,1H3

InChI key

KDOUBUZYGVSSLM-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/5-fluoro-2-methylbenzylamine

Synonym

(5-Fluoro-2-methylphenyl)methylamine, (5-Fluoro-o-tolyl)methylamine

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/5-fluoro-2-methylbenzylamine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service