Skip to Content
Merck
All Photos(1)

Key Documents

APO455832619

Apollo Scientific Ltd

4-[2-(Piperazin-1-yl)ethyl]morpholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 59.80

Vendor SKU: OR6912-1G

CHF 59.80


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 59.80

About This Item

CAS Number:
Molecular Weight:
199.3
MDL number:

Vendor SKU: OR6912-1G

CHF 59.80


Ships in 14 days - from Aldrich Partner.

Iupac Name

4-[2-(1-piperazinyl)ethyl]morpholine

Inchi Code

1S/C10H21N3O/c1-3-12(4-2-11-1)5-6-13-7-9-14-10-8-13/h11H,1-10H2

InChI key

SAJZEJMFAWZNCQ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid-Viscous

Product Link

https://store.apolloscientific.co.uk/product/4-2-piperazin-1-ylethylmorpholine

Flash Point

106-107/0.2mm

Pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-2-piperazin-1-ylethylmorpholine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service