Skip to Content
Merck
All Photos(1)

Key Documents

APO443485371

Apollo Scientific Ltd

4-Bromo-5-chloro-1H-benzimidazole

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 318.30
1 G
CHF 915.60

Vendor SKU: OR400381-250MG

CHF 318.30


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CHF 318.30
1 G
CHF 915.60

About This Item

CAS Number:
Molecular Weight:
231.48
MDL number:

Vendor SKU: OR400381-250MG

CHF 318.30


Ships in 14 days - from Aldrich Partner.

Iupac Name

4-bromo-5-chloro-1H-benzo[d]imidazole

Inchi Code

1S/C7H4BrClN2/c8-6-4(9)1-2-5-7(6)11-3-10-5/h1-3H,(H,10,11)

InChI key

PQDFVUZVPCDZFJ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/4-bromo-5-chloro-1h-benzimidazole

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-bromo-5-chloro-1h-benzimidazole


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service