Skip to Content
Merck
All Photos(1)

Key Documents

APO428499410

Apollo Scientific Ltd

4-Ethoxy-2,6-difluorobenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 92.80
5 G
CHF 133.50
25 G
CHF 352.40

Vendor SKU: PC302746-1G

CHF 92.80


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 92.80
5 G
CHF 133.50
25 G
CHF 352.40

About This Item

CAS Number:
Molecular Weight:
186.16
MDL number:

Vendor SKU: PC302746-1G

CHF 92.80


Ships in 14 days - from Aldrich Partner.

bp

70-72

Iupac Name

4-ethoxy-2,6-difluorobenzaldehyde

Inchi Code

1S/C9H8F2O2/c1-2-13-6-3-8(10)7(5-12)9(11)4-6/h3-5H,2H2,1H3

InChI key

SHCJRMIWXNHWJF-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/4-ethoxy-26-difluorobenzaldehyde

Boiling point

70-72

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-ethoxy-26-difluorobenzaldehyde


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service