Skip to Content
Merck
All Photos(1)

Key Documents

AOBH9910CC68

AOBChem

2-fluoro-3-(hydroxymethyl)phenol

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 210.64
500 MG
CHF 275.56
1 G
CHF 361.29

Vendor SKU: 118029-250MG

CHF 210.64


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CHF 210.64
500 MG
CHF 275.56
1 G
CHF 361.29

About This Item

CAS Number:
Molecular Weight:
142.13
MDL number:

Vendor SKU: 118029-250MG

CHF 210.64


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-fluoro-3-(hydroxymethyl)phenol

Inchi Code

1S/C7H7FO2/c8-7-5(4-9)2-1-3-6(7)10/h1-3,9-10H,4H2

InChI key

ZFMHUICCOQUFDS-UHFFFAOYSA-N

Purity

95

Country of Origin

CN

Physical Form

Solid

Product Link

https://aobchem.com/2-fluoro-3-hydroxymethyl-phenol-118029.html

MSDS

https://aobchem.com/pub/media/files/sds/118029.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service