Skip to Content
Merck
All Photos(1)

Key Documents

AOB640095266

AOBChem

2,4-dichloro-1-isopropoxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 132.26
1 G
CHF 161.65
5 G
CHF 426.20
10 G
CHF 682.17

Vendor SKU: 24328-500MG

CHF 132.26


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
500 MG
CHF 132.26
1 G
CHF 161.65
5 G
CHF 426.20
10 G
CHF 682.17

About This Item

CAS Number:
Molecular Weight:
205.08
MDL number:

Vendor SKU: 24328-500MG

CHF 132.26


Ships in 14 days - from Aldrich Partner.

Iupac Name

2,4-dichlorophenyl isopropyl ether

Inchi Code

1S/C9H10Cl2O/c1-6(2)12-9-4-3-7(10)5-8(9)11/h3-6H,1-2H3

InChI key

SKZUJEUNTYZIRO-UHFFFAOYSA-N

Purity

95

Country of Origin

CN

Physical Form

Liquid

Product Link

https://aobchem.com/2-4-dichloro-1-isopropoxybenzene.html

MSDS

https://aobchem.com/pub/media/files/sds/24328.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service