WHA77073800
Whatman® 850-DS 8-channel filter plate
PVDF membrane, pore size 0.45 μm, pack of 50 ea
Synonym(s):
8-channel Filter Plate, 850-DS Filter Plate
Select a Size
CHF 537.00
Estimated to ship on14 April 2025
Select a Size
About This Item
CHF 537.00
Estimated to ship on14 April 2025
Recommended Products
material
PVDF membrane
packaging
pack of 50 ea
manufacturer/tradename
Cytiva
pore size
0.45 μm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Automated processing: up to 8 samples simultaneously
- Readily available: in a wide range of pore sizes and materials
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service