SLWSST102K
Pyrex® Spare PTFE keys for Pyrex® Stopcocks
straight key bore, bore 1.5 mm , key
Synonym(s):
Pyrex, SciLabware SST102K
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
SST102K
SciLabware
bore
1.5 mm , key
stopcock diam.
12.5 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Fully interchangeable PTFE keys for Pyrex stopcocks
- Supplied complete with plastic screw thread retaining device
- Keys are self lubricating, avoiding the need for stopcock grease
- Made to a taper of 1:10 giving a combination of performance and ease of operation
Features and Benefits
• PTFE stopcock key with silicone ′O′ ring and PP retaining device• PTFE keys are fully interchangeable• Keys are self lubricating, avoiding the need for stopcock grease• Keys have a 1:10 taper to accurately fit the relevant glass stopcock barrel
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service