Key Documents
SLW4503/22
Pyrex® Culture tube caps with insert
thread size 22 mm
Synonym(s):
Pyrex, SciLabware 4503/22
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
thread size
22 mm
manufacturer/tradename
4503/22
SciLabware
General description
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today′s research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used.
Features and Benefits
• Manufactured from borosilicate glass 3.3• High resistance to attack from water reduces leaching of contaminants which can cause pH changes• PBT screwcaps with inert PTFE lined rubber disc• Cap can withstand repeated dry heat and steam sterilisation cycles.
Other Notes
Cap and insert only.
Legal Information
PYREX is a registered trademark of Corning, Inc.