GF97173153
Aluminum-silicon alloy, Al96Si4
powder, 150micron max. particle size, 50g
Synonym(s):
AB036010
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
powder (atomized)
manufacturer/tradename
Goodfellow 971-731-53
weight
50 g
max. part. size
150 μm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service