Skip to Content
Merck
All Photos(2)

Documents

DWK218065435

Duran® Gl 45 Laboratory Glass Bottle Protect

plastic-coated glass bottle (PU), amber bottle, with screw cap to drain contents: no (Without pouring ring)

Synonym(s):

218065435

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

amber bottle
plastic-coated glass bottle (PU)

feature

with screw cap to drain contents: no (Without pouring ring)

packaging

pkg of 10 ea

manufacturer/tradename

DURAN® 218065435

Looking for similar products? Visit Product Comparison Guide

Application

Preparation of light-sensitive materials, Storage of light-sensitive materials, Transport and shipping of light-sensitive materials, Autoclaving of media and reagents, Culturing vessel, Boro 3.3, plastic coated

Features and Benefits

Standardised GL-thread and matching cap system for practicularly tight sealing and simple, clean pouring out; UV protection up to approx. 500 nm due to external colouration; The only external application of the colour ensures that internal surfaces have standard DURAN® glass properties; plastic coatedVery durable and chemically resistant amber colour ; Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working; Easy-to-read scale with graduation marks to estimate the content; Fired-on, highly durable white ceramic print; Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service