All Photos(1)
About This Item
Linear Formula:
ClCH=CHCOOH
CAS Number:
Molecular Weight:
106.51
Beilstein:
1720279
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
Assay
99%
SMILES string
OC(=O)\C=C\Cl
InChI
1S/C3H3ClO2/c4-2-1-3(5)6/h1-2H,(H,5,6)/b2-1+
InChI key
MHMUCYJKZUZMNJ-OWOJBTEDSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service