Skip to Content
Merck
All Photos(2)

Key Documents

911615

Sigma-Aldrich

[Ru(η33-C10H16)Cl(μ2-O,O-O2CMe)]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19ClO2Ru
CAS Number:
Molecular Weight:
331.80
UNSPSC Code:
12161600

form

powder or crystals

mp

141-147 °C

Application

[Ru(η33-C10H16)Cl(μ2-O,O-O2CMe)] is a ruthenium catalyst developed for the redox-neutral isomerization of allylic alcohols to saturated carbonyls under physiological conditions. This bioorthoganol and biocompatible catalyst has also been shown to be active inside living cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolás Ríos-Lombardía et al.
Chemical communications (Cambridge, England), 51(54), 10937-10940 (2015-06-13)
The ruthenium-catalysed isomerisation of allylic alcohols was coupled, for the first time, with asymmetric bioamination in a one-pot process in an aqueous medium. In the cases involving prochiral ketones, the ω-TA exhibited excellent enantioselectivity, identical to that observed in the
Cristian Vidal et al.
Journal of the American Chemical Society, 141(13), 5125-5129 (2019-03-21)
Tailored ruthenium(IV) complexes can catalyze the isomerization of allylic alcohols into saturated carbonyl derivatives under physiologically relevant conditions, and even inside living mammalian cells. The reaction, which involves ruthenium-hydride intermediates, is bioorthogonal and biocompatible, and can be used for the
Nicolás Ríos-Lombardía et al.
Angewandte Chemie (International ed. in English), 55(30), 8691-8695 (2016-06-04)
The ruthenium-catalyzed redox isomerization of allylic alcohols was successfully coupled with the enantioselective enzymatic ketone reduction (mediated by KREDs) in a concurrent process in aqueous medium. The overall transformation, formally the asymmetric reduction of allylic alcohols, took place with excellent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service