Skip to Content
Merck
All Photos(2)

Documents

901250

Sigma-Aldrich

2-(Pyridin-2-yl)quinolin-8-amine dihydrochloride

Synonym(s):

PAQ directing group

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N3 · 2HCl
Molecular Weight:
294.18
UNSPSC Code:
12352101

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

Application

Developed in the Engle lab, this 2-pyridyl-8-aminoquinoline (PAQ) auxiliary is a pincer-like, removable tridentate directing group that stabilizes 6-membered palladacycles for olefin functionalization. Together with similar ligand PPA (901251), researchers demonstrated regioselective remote hydrocarbofunctionalization of alkene-containing substrate classes (e.g. 4-pentenoic acids, allylic alcohols, homoallyl amines, bis-homoallylamines) using Pd(II) catalysis.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miriam L O'Duill et al.
Journal of the American Chemical Society, 139(44), 15576-15579 (2017-10-04)
Removable tridentate directing groups inspired by pincer ligands have been designed to stabilize otherwise kinetically and thermodynamically disfavored 6-membered alkyl palladacycle intermediates. This family of directing groups enables regioselective remote hydrocarbofunctionalization of several synthetically useful alkene-containing substrate classes, including 4-pentenoic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service