Skip to Content
Merck
All Photos(1)

Documents

738123

Sigma-Aldrich

2,6-Dichloro-1-fluoropyridinium triflate

95%

Synonym(s):

1-Fluoro-2,6-dichloropyridinium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl2F4NO3S
CAS Number:
Molecular Weight:
316.06
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

121-126 °C

storage temp.

2-8°C

SMILES string

[O-]S(=O)(=O)C(F)(F)F.F[n+]1c(Cl)cccc1Cl

InChI

1S/C5H3Cl2FN.CHF3O3S/c6-4-2-1-3-5(7)9(4)8;2-1(3,4)8(5,6)7/h1-3H;(H,5,6,7)/q+1;/p-1

InChI key

KDFRVNARKRKXQQ-UHFFFAOYSA-M

Application

  • Used in the fluorination of various organic compounds
  • Fluoropyridinium triflates are versatile reagents for transformation of thioglycosides into ο-glycoside, glycosyl azide and sulfoxide
  • Electrolytic partial fluorination and diastereoselective anodic fluorination
  • Regioselective, electrochemical fluorination
  • Fluorination of alkenes with N-F type of reagents

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrolytic partial fluorination of organic compound. Part: 53 Highly regioselective anodic mono- and difluorination of 4-arylthio-1,3-dioxolan-2-ones. A marked solvent effect on fluorinated product selectivity
Ishii, H.; et al.
Tetrahedron, 57, 9067-9072 (2001)
Electrolytic partial fluorination of organic compounds. Part 62. Highly diastereoselective anodic fluorination of chiral 1,3-oxathiolan-5-ones derived from camphorsulfonamides
Baba, D.; et al.
Journal of Fluorine Chemistry, 121, 93-96 (2003)
1-Fluoropyridinium triflates: versatile reagents for transformation of thioglycoside into O-glycoside, glycosyl azide and sulfoxide
Tsukamoto, H.; et al.
Tetrahedron Letters, 44, 5247-5249 (2003)
Power- and structure-variable fluorinating agents. The N-fluoropyridinium salt system
Umemoto, T.; et al.
Journal of the American Chemical Society, 112, 8563-8575 (1990)
Evidence for ionic intermediates in fluorination of alkenes with N-F type of reagents
Zupan, M.; et al.
Chemistry Letters (Jpn), 7, 641-642 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service