Skip to Content
Merck
All Photos(1)

Key Documents

708275

Sigma-Aldrich

1,3-Bis-(2,6-diisopropylphenyl)-[1,3,2]diazaphospholidine 2-oxide

97%

Synonym(s):

1,3-Bis[2,6-bis(1-methylethyl)phenyl]-1,3,2-diazaphospholidine 2-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H39N2OP
CAS Number:
Molecular Weight:
426.57
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

205-210 °C

functional group

phosphine

SMILES string

CC(C)c1cccc(C(C)C)c1N2CCN(c3c(cccc3C(C)C)C(C)C)[PH]2=O

InChI

1S/C26H39N2OP/c1-17(2)21-11-9-12-22(18(3)4)25(21)27-15-16-28(30(27)29)26-23(19(5)6)13-10-14-24(26)20(7)8/h9-14,17-20,30H,15-16H2,1-8H3

InChI key

SQYNIVDXVZPMLW-UHFFFAOYSA-N

Application

Ligand used with ruthenium for dehydrative direct arylation of arenes with phenols[1]
Reactnat for synthesis of air-stable palladium HASPO complexes

Catalyst for:
  • Dehydrative direct arylations of arenes
  • Aryl-(hetero)aryl coupling by activation of C-Cl and C-F bonds

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lutz Ackermann et al.
Organic letters, 10(21), 5043-5045 (2008-10-16)
Phenols can be employed as proelectrophiles in operationally simple ruthenium-catalyzed dehydrative direct arylations, proceeding through chemo- and regioselective functionalizations of C-H and C-OH bonds.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service