Skip to Content
Merck
All Photos(2)

Key Documents

641855

Sigma-Aldrich

Samarium(III) oxide dispersion

nanoparticles, <100 nm particle size (BET)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sm2O3
CAS Number:
Molecular Weight:
348.72
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

liquid

composition

H2O (Solvent)

reaction suitability

reagent type: catalyst
core: samarium

particle size

<100 nm (BET)
<50 nm (XRD)

SMILES string

O=[Sm]O[Sm]=O

InChI

1S/3O.2Sm

InChI key

PRCWVHVINXAFRG-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H M T U Herath et al.
Journal of biomedical materials research. Part A, 94(1), 130-136 (2010-02-04)
The biocompatibility of natural samarium (III) oxide, which has previously been used for treatment in bone-related diseases was determined as a first step in its evaluation as a bone implant material. Assessment for 28 days using osteoblast-like cells revealed no
Guohui Zhang et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 67(1), 46-49 (2008-08-19)
Cross sections of the (147)Sm(n, alpha)(144)Nd reaction were measured at En=5.0 and 6.0MeV. A twin gridded ionization chamber was used as a charged particle detector and two large area (147)Sm(2)O(3) samples placed back to back were employed. Experiments were performed
Janne Marvola et al.
International journal of pharmaceutics, 281(1-2), 3-10 (2004-08-04)
This paper is a report from a pharmacoscintigraphic study with an Egalet constant-release system containing caffeine and natural abundance samarium oxide. First the formulation was tested in vitro to clarify integrity during irradiation in the nuclear reactor. Then six healthy
B Mandal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(4), 933-939 (2004-03-24)
Microphone based photoacoustic (PA) spectrometer to study solids in powder form was designed and fabricated. Laser PA spectra of Sm3+ ion in Sm2O3 and SmCl3.6H2O microcrystalline powders were recorded first time in 484-542 nm spectral region at room temperature. Analysis
Matthew D Burke et al.
Pharmaceutical research, 24(4), 695-704 (2007-03-21)
To develop a robust radiolabeling technique to enable evaluation of difficult to radiolabel gastric retentive formulations using gamma scintigraphy. The use of a successful radiolabel will allow accurate assessment of the gastric residence time of the formulations. The retention of

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service