Skip to Content
Merck
All Photos(2)

Key Documents

637327

Sigma-Aldrich

Holmium(III) oxide

nanopowder, <100 nm avg. part. size (DLS), ≥99.9% trace metals basis

Synonym(s):

Rare earth

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
CHF 221.00

CHF 221.00


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
10 G
CHF 221.00

About This Item

Linear Formula:
Ho2O3
CAS Number:
Molecular Weight:
377.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

CHF 221.00


Estimated to ship on24 April 2025


Request a Bulk Order

Quality Level

Assay

≥99.9% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: holmium

avg. part. size

<100 nm (DLS)

particle size

<50 nm (XRD)

mp

2330 °C (lit.)

SMILES string

O=[Ho]O[Ho]=O

InChI

1S/2Ho.3O

InChI key

JYTUFVYWTIKZGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service