Skip to Content
Merck
All Photos(1)

Documents

495581

Sigma-Aldrich

3-Phenoxyphenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4NCO
CAS Number:
Molecular Weight:
211.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.5925 (lit.)

bp

294 °C (lit.)

density

1.17 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=Nc1cccc(Oc2ccccc2)c1

InChI

1S/C13H9NO2/c15-10-14-11-5-4-8-13(9-11)16-12-6-2-1-3-7-12/h1-9H

InChI key

HOJSXCMKZBXNEN-UHFFFAOYSA-N

General description

3-Phenoxyphenyl isocyanate, also known as 1-isocyanato-3-phenoxybenzene, is an organic building block containing an isocyanate group.

Application

3-Phenoxyphenyl isocyanate may be used in the synthesis of 1-(8-chloro-[1,2,4]triazolo[4,3-a]pyridine-3-carbonyl)-4-(3-phenoxyphenyl)semicarbazide.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-assisted synthesis of novel 8-chloro-[1,2,4] triazolo [4,3-a] pyridine derivatives.
Zhang L, et al.
Turkish Journal of Chemistry, 39(4), 867-873 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service